missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals Product Code.: 11436907

2-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals

Product Code. 11436907
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11436907

Brand: Thermo Scientific Alfa Aesar A14343.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson's reagent and solvent tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson′s reagent and solvent tetrahydrofuran.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-32-1
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00045170
InChI Key WGNNILPYHCKCFF-UHFFFAOYSA-N
Synonym n-methyl-2-chloroaniline, benzenamine, 2-chloro-n-methyl, n1-methyl-2-chloroaniline, 2-chloro-n-methylbenzenamine, o-chlormonomethylanilin, 2-chlor-n-methylaniline, 2-chloro-n-methyl aniline, 2-chloro-n-methyl-aniline, 2-methylamino chlorobenzene, 2-chloranyl-n-methyl-aniline
PubChem CID 136736
IUPAC Name 2-chloro-N-methylaniline
SMILES CNC1=CC=CC=C1Cl

Specifications

Density 1.15
Boiling Point 218°C
Flash Point 92°C (197°F)
Refractive Index 1.58
Quantity 1 g
UN Number UN2810
Beilstein 2802444
Solubility Information Not miscible or difficult to mix.
Formula Weight 141.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-methylaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.