missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N,N-dimethyl-4-nitroaniline, 96%, Thermo Scientific™

Product Code. 15450209
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6213-19-0
Molecular Formula C8H9ClN2O2
Molecular Weight (g/mol) 200.622
MDL Number MFCD00043604
InChI Key OZKAWTHGBGLZKC-UHFFFAOYSA-N
Synonym 2-chloro-nn-dimethyl-4-nitroaniline, benzenamine, 2-chloro-n,n-dimethyl-4-nitro, n,n-dimethyl-4-nitro-2-chloroaniline, 2-chloro-4-nitrophenyl dimethylamine, 3-chloro-4-dimethylamino nitrobenzene, 3-chloro-4-dimethylaminonitrobenzene, 2-chloro-n,n-dimethyl-4-nitrobenzenamine, benzenamine,2-chloro-n,n-dimethyl-4-nitro, 2-chloro-4-nitro-n,n-dimethylaniline
PubChem CID 80343
IUPAC Name 2-chloro-N,N-dimethyl-4-nitroaniline
SMILES CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])Cl

Specifications

Quantity 250 mg
UN Number UN2811
Formula Weight 200.62
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Chloro-N,N-dimethyl-4-nitroaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'