missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloroaniline, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -3°C |
CAS Min % | 98.0 |
Color | Yellow to Brown |
CAS Max % | 100.0 |
Density | 1.2100g/mL |
Boiling Point | 209°C |
Quantity | 250 mL |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Physical Form | Liquid |
Show More |
Safety and Handling
Product Identifier
- 2-Chloroaniline
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'