missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzophenone, 99+%, Thermo Scientific Chemicals

Product Code. 11492568
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5162-03-8
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.66
MDL Number MFCD00000558
InChI Key VMHYWKBKHMYRNF-UHFFFAOYSA-N

Specifications

Melting Point 44°C to 47°C
Boiling Point 330°C
Flash Point >110°C (230°F)
Quantity 500 g
Beilstein 1869594
Synonym sodium thiomethoxide, sodium methanethiolate, sodium thiomethylate, methyl mercaptan sodium salt, methanethiol sodium salt, sodiumthiomethoxide, methanethiol, sodium salt, sodiummethanethiolate, sodium methyl mercaptide, methylsulfanyl sodium
SMILES ClC1=CC=CC=C1C(=O)C1=CC=CC=C1
IUPAC Name (2-chlorophenyl)(phenyl)methanone
PubChem CID 4378561
Formula Weight 216.67
Percent Purity ≥99%
Chemical Name or Material 2-Chlorobenzophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'