missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzotrifluoride, 99%

Product Code. 11409642
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11409642

Brand: Thermo Scientific Alfa Aesar A10753.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-16-4 | C7H4ClF3 | 180.554 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-16-4
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.554
MDL Number MFCD00000561
InChI Key DGRVQOKCSKDWIH-UHFFFAOYSA-N
Synonym 2-chlorobenzotrifluoride, 1-chloro-2-trifluoromethyl benzene, o-chlorobenzotrifluoride, chlorobenzotrifluoride, benzene, 1-chloro-2-trifluoromethyl, 2-chloro trifluoromethyl benzene, 2-chloro-alpha,alpha,alpha-trifluorotoluene, 2-chloro-benzotrifluoride, o-trifluoromethyl chlorobenzene, o-trifluoromethyl phenyl chloride
PubChem CID 6921
IUPAC Name 1-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)Cl

Specifications

Melting Point -6°C
Density 1.369
Boiling Point 151°C to 153°C
Flash Point 58°C (136°F)
Refractive Index 1.456
Quantity 100 g
UN Number UN2234
Beilstein 510993
Formula Weight 180.56
Percent Purity 99%
Chemical Name or Material 2-Chlorobenzotrifluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chlorobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Chlorobenzotrifluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.