missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzotrifluoride, 99%, Thermo Scientific Chemicals

Product Code. 11419642
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-16-4
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.554
MDL Number MFCD00000561
InChI Key DGRVQOKCSKDWIH-UHFFFAOYSA-N
Synonym 2-chlorobenzotrifluoride, 1-chloro-2-trifluoromethyl benzene, o-chlorobenzotrifluoride, chlorobenzotrifluoride, benzene, 1-chloro-2-trifluoromethyl, 2-chloro trifluoromethyl benzene, 2-chloro-alpha,alpha,alpha-trifluorotoluene, 2-chloro-benzotrifluoride, o-trifluoromethyl chlorobenzene, o-trifluoromethyl phenyl chloride
PubChem CID 6921
IUPAC Name 1-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)Cl

Specifications

Melting Point -6°C
Density 1.369
Boiling Point 151°C to 153°C
Flash Point 58°C (136°F)
Refractive Index 1.456
Quantity 500 g
UN Number UN2234
Beilstein 510993
Formula Weight 180.56
Percent Purity 99%
Chemical Name or Material 2-Chlorobenzotrifluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'