missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11472507
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

Solubility
Soluble in acetone, ether, and alcohol. Solubility in water it decomposes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong bases, alcohols, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-65-4
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00000660
InChI Key ONIKNECPXCLUHT-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-chloro, o-chlorobenzoyl chloride, chlorobenzoyl chloride, benzoyl chloride, o-chloro, ococ, 2-chloro-benzoyl chloride, unii-jr29a4n74x, o-chlorobenzoylchloride, 2-chlorobenzoylchloride, 2-chlorobenzoic acid chloride
PubChem CID 69110
ChEBI CHEBI:60719
IUPAC Name 2-chlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC=C1Cl

Specifications

Melting Point -4°C
Density 1.379
Boiling Point 237°C to 239°C
Flash Point 125°C (257°F)
Odor Acrid
Refractive Index 1.5718
Quantity 1000 g
UN Number UN3265
Beilstein 386435
Sensitivity Moisture sensitive
Solubility Information Soluble in acetone,ether,and alcohol. Solubility in water it decomposes.
Formula Weight 175.01
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'