missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroethanol, 99+%, Pure, Fisher Chemical
Click to view available options
500mL, Plastic Coated Glass bottle
Specifications
CAS | 107-07-3 |
Melting Point | -63°C |
Boiling Point | 129°C |
Packaging | Plastic Coated Glass bottle |
Molecular Formula | C2H5ClO |
MDL Number | 2829 |
Quantity | 500 mL |
Formula Weight | 80.51g/mol |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'