missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroethyl methyl ether, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | -55°C |
Color | Colorless to Yellow |
Density | 1.0300g/mL |
Boiling Point | 89°C to 90°C |
Flash Point | 15°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- 2-Chloroethyl methyl ether
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 2
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'