missing translation for 'onlineSavingsMsg'
Learn More

2-chloromandelic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10740765
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10421-85-9
C8H7ClO3
186.59
MFCD00084962
RWOLDZZTBNYTMS-UHFFFAOYNA-N
2-chloromandelic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, dl-2-chloromandelic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, 2-2-chlorophenyl-2-hydroxy-acetic acid, 2-chlorophenyl glycolic acid, 2-chloromandelicacid, 2-chloro-mandelic acid, pubchem15895
97720
2-(2-chlorophenyl)-2-hydroxyacetic acid
OC(C(O)=O)C1=CC=CC=C1Cl
This item is not returnable. View return policy
10421-85-9
C8H7ClO3
186.59
MFCD00084962
RWOLDZZTBNYTMS-UHFFFAOYNA-N
2-chloromandelic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, dl-2-chloromandelic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, 2-2-chlorophenyl-2-hydroxy-acetic acid, 2-chlorophenyl glycolic acid, 2-chloromandelicacid, 2-chloro-mandelic acid, pubchem15895
97720
2-(2-chlorophenyl)-2-hydroxyacetic acid
OC(C(O)=O)C1=CC=CC=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 80°C to 85°C
CAS Min % 97.5
Color Cream to White
CAS Max % 100.0
Quantity 250 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 97.5% min. (HPLC)
Linear Formula ClC6H4CH(OH)COOH
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 2-Chloromandelic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloromandelic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'