Learn More
2-Chlorophenol, 98+%, Thermo Scientific Chemicals
£27.54 - £381.90
Chemical Identifiers
CAS | 95-57-8 |
---|---|
Molecular Formula | C6H5ClO |
Molecular Weight (g/mol) | 128.56 |
MDL Number | MFCD00002159 |
InChI Key | ISPYQTSUDJAMAB-UHFFFAOYSA-N |
Synonym | o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol |
PubChem CID | 7245 |
ChEBI | CHEBI:47083 |
IUPAC Name | 2-chlorophenol |
SMILES | C1=CC=C(C(=C1)O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10722231
|
Thermo Scientific Acros
180990050 |
5 g | Glass bottle |
£27.54
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10072250
|
Thermo Scientific Acros
180991000 |
100 g | Glass bottle |
£31.96
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10635632
|
Thermo Scientific Acros
180995000 |
500 g | Glass bottle |
£45.83
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10239603
|
Thermo Scientific Acros
180990025 |
2.5 kg | Glass bottle |
£142.71
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11954681
|
Thermo Scientific Acros
180990100 |
10 kg | Plastic Can |
£381.90
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-57-8 | |
128.56 | |
ISPYQTSUDJAMAB-UHFFFAOYSA-N | |
7245 | |
2-chlorophenol |
C6H5ClO | |
MFCD00002159 | |
o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol | |
CHEBI:47083 | |
C1=CC=C(C(=C1)O)Cl |
Specifications
95-57-8 | |
100.0 | |
1.2620g/mL | |
63°C | |
C6H5ClO | |
1.5580 to 1.5600 | |
06,183 | |
1.262 | |
Solubility in water: 28.5g/l (20°C). Other solubilities: freely soluble in solutions of sodium hydroxide,, alcohol and ether, soluble in aromatic solvents | |
C1=CC=C(C(=C1)O)Cl | |
128.56 | |
CHEBI:47083 | |
128.56 | |
2-Chlorophenol, 98+% |
98.0 | |
8.0°C | |
173.0°C to 175.0°C | |
98% min. (GC) | |
ClC6H4OH | |
MFCD00002159 | |
15,2157 | |
o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol | |
ISPYQTSUDJAMAB-UHFFFAOYSA-N | |
2-chlorophenol | |
7245 | |
4.11 mPa.s (25°C) | |
98+% |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-433-2
missing translation for 'rtecsNumber' : SK2625000
missing translation for 'tsca' : TSCA
RUO – Research Use Only