missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylacetic acid, 98.5+%, Thermo Scientific Chemicals

Product Code. 10168123
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2444-36-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
MDL Number MFCD00004317
InChI Key IUJAAIZKRJJZGQ-UHFFFAOYSA-N
Synonym 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid
PubChem CID 17124
IUPAC Name 2-(2-chlorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)Cl

Specifications

Melting Point 94.0°C to 97.0°C
CAS Min % 98.5
Color White to Yellow
CAS Max % 100.0
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Assay Percent Range 98.5+%
Linear Formula ClC6H4CH2CO2H
Beilstein 09, 447
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 170.6
Percent Purity 98.5+%
Chemical Name or Material 2-Chlorophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'