Learn More
2-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
£21.20 - £61.20
Chemical Identifiers
CAS | 41052-75-9 |
---|---|
Molecular Formula | C6H8Cl2N2 |
Molecular Weight (g/mol) | 179.04 |
MDL Number | MFCD00012928 |
InChI Key | ADODRSVGNHNKAT-UHFFFAOYSA-N |
Synonym | 2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride |
PubChem CID | 443287 |
ChEBI | CHEBI:1046 |
IUPAC Name | (2-chlorophenyl)hydrazine;hydrochloride |
SMILES | [H+].[Cl-].NNC1=CC=CC=C1Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10133340
|
Thermo Scientific Acros
149140050 |
5 g | Glass bottle |
£21.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10563501
|
Thermo Scientific Acros
149140250 |
25 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
41052-75-9 | |
179.04 | |
ADODRSVGNHNKAT-UHFFFAOYSA-N | |
443287 | |
(2-chlorophenyl)hydrazine;hydrochloride |
C6H8Cl2N2 | |
MFCD00012928 | |
2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride | |
CHEBI:1046 | |
[H+].[Cl-].NNC1=CC=CC=C1Cl |
Specifications
41052-75-9 | |
100.0 | |
White | |
96% min. (Argentometry) | |
ClC6H4NHNH2·HCl | |
15, 422 | |
Solubility in water: soluble | |
[H+].[Cl-].NNC1=CC=CC=C1Cl | |
179.04 | |
CHEBI:1046 | |
97% | |
2-Chlorophenylhydrazine hydrochloride, 97% |
96.0 | |
186°C to 188°C | |
Authentic | |
C6H8Cl2N2 | |
MFCD00012928 | |
2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride | |
ADODRSVGNHNKAT-UHFFFAOYSA-N | |
(2-chlorophenyl)hydrazine;hydrochloride | |
443287 | |
179.05 | |
Flakes |
Safety and Handling
GHS H Statement
Very toxic to aquatic life.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Do not eat,drink or smoke when using this product.
IF
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 255-194-1
missing translation for 'tsca' : TSCA
RUO – Research Use Only