Learn More
2-chloropyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
£21.20 - £68.00
Chemical Identifiers
CAS | 452972-11-1 |
---|---|
Molecular Formula | C11H15BClNO2 |
Molecular Weight (g/mol) | 239.51 |
MDL Number | MFCD04039873 |
InChI Key | XFZFMAUUZHBQSS-UHFFFAOYSA-N |
Synonym | 2-chloropyridine-3-boronic acid pinacol ester, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-chloro-3-pyridineboronic acid pinacol ester, 2-chloropyridin-3-ylboronic acid pinacol ester, 2-chloropyridine-3-boronicacidpinacolester, 2-chloropyridine-3-boronic acid, pinacol ester, 2-chloro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem16529 |
PubChem CID | 12067060 |
IUPAC Name | 2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
SMILES | CC1(C)OB(OC1(C)C)C1=CC=CN=C1Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10778488
|
Thermo Scientific Acros
430660010 |
1 g | Glass bottle | ||||||
10504418
|
Thermo Scientific Acros
430660050 |
5 g | Glass bottle |
£68.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
452972-11-1 | |
239.51 | |
XFZFMAUUZHBQSS-UHFFFAOYSA-N | |
12067060 | |
CC1(C)OB(OC1(C)C)C1=CC=CN=C1Cl |
C11H15BClNO2 | |
MFCD04039873 | |
2-chloropyridine-3-boronic acid pinacol ester, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-chloro-3-pyridineboronic acid pinacol ester, 2-chloropyridin-3-ylboronic acid pinacol ester, 2-chloropyridine-3-boronicacidpinacolester, 2-chloropyridine-3-boronic acid, pinacol ester, 2-chloro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem16529 | |
2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Specifications
452972-11-1 | |
100.0 | |
Brown to White | |
96% min. (GC) | |
MFCD04039873 | |
XFZFMAUUZHBQSS-UHFFFAOYSA-N | |
2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
12067060 | |
97% | |
2-chloropyridine-3-boronic acid pinacol ester |
96.0 | |
34.0°C to 39.0°C | |
Authentic | |
C11H15BClNO2 | |
2-chloropyridine-3-boronic acid pinacol ester, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-chloro-3-pyridineboronic acid pinacol ester, 2-chloropyridin-3-ylboronic acid pinacol ester, 2-chloropyridine-3-boronicacidpinacolester, 2-chloropyridine-3-boronic acid, pinacol ester, 2-chloro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem16529 | |
CC1(C)OB(OC1(C)C)C1=CC=CN=C1Cl | |
239.51 | |
239.61 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
- 2-Chloropyridine-3-boronic acid pinacol ester
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only