missing translation for 'onlineSavingsMsg'
Learn More
2-Chloropyridine, 99%, Thermo Scientific Chemicals Product Code.: 10624762

2-Chloropyridine, 99%, Thermo Scientific Chemicals

Product Code. 10624762
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10624762

Brand: Thermo Scientific Acros 110011000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-09-1
MDL Number MFCD00006228
InChI Key OKDGRDCXVWSXDC-UHFFFAOYSA-N
Synonym o-chloropyridine, pyridine, 2-chloro, alpha-chloropyridine, .alpha.-chloropyridine, unii-8hmd45ayej, 2-chloro-pyridine, ccris 1724, 8hmd45ayej, 6-chloro-pyridine, dsstox_cid_4810
PubChem CID 7977
ChEBI CHEBI:39174
IUPAC Name 2-chloropyridine
SMILES C1=CC=NC(=C1)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -46.0°C
Color Colorless to Yellow
Density 1.2090g/mL
Boiling Point 168.0°C to 170.0°C
Flash Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H4ClN
Linear Formula (C5H4N)Cl
Refractive Index 1.5310 to 1.5330
Quantity 100 mL
Beilstein 20, 230
Specific Gravity 1.209
Solubility Information Solubility in water: 27g/L (20°C). Other solubilities: >50g/100mL ethanol,soluble in ether and alcohol
Molecular Weight (g/mol) 113.55
Formula Weight 113.55
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloropyridine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-Chloropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloropyridine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.