missing translation for 'onlineSavingsMsg'
Learn More

2-chloropyrimidine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11956541
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 933702-55-7
Molecular Formula C5H3ClN2O
Molecular Weight (g/mol) 142.54
MDL Number MFCD10696891
InChI Key LJYQVOPFBNMTKJ-UHFFFAOYSA-N
Synonym 2-chloropyrimidine-5-carboxyaldehyde, 2-chloropyrimidine-5-carboxaldehyde, 2-chloro-5-pyrimidinecarboxaldehyde, pubchem23896, 2-chloro-5-formyl pyrimidine, chloropyrimidine-5-carbaldehyde, 2-chloro-pyrimidine-5-carbaldehyde, 2-chloranylpyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde, 2-chloro
PubChem CID 21698350
IUPAC Name 2-chloropyrimidine-5-carbaldehyde
SMILES ClC1=NC=C(C=O)C=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 113°C to 118°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 142.55
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Chloropyrimidine-5-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloropyrimidine-5-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/physician/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'