missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 11309639
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4?-(1,1?-(5-(2-methoxyphenoxy)-[2,2?-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4′-(1,1′-(5-(2-methoxyphenoxy)-[2,2′-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

Solubility
Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1722-12-9
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD00006060
InChI Key UNCQVRBWJWWJBF-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
PubChem CID 74404
IUPAC Name 2-chloropyrimidine
SMILES ClC1=NC=CC=N1

Specifications

Melting Point 64°C to 67°C
Boiling Point 75°C to 76°C (10 mmHg)
Flash Point 98°C (208°F)
Quantity 10 g
UN Number UN2811
Beilstein 107171
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol.
Formula Weight 114.54
Percent Purity 98%
Chemical Name or Material 2-Chloropyrimidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'