missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyano-4'-methylbiphenyl, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 49°C to 53°C |
Density | 1.17 |
Boiling Point | >320°C |
Quantity | 5 g |
UN Number | UN3077 |
Beilstein | 3605954 |
Formula Weight | 193.26 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Cyano-4'-methylbiphenyl |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H302-Harmful if swallowed.
- H361-Suspected of damaging fertility or the unborn child.
- H372-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'