missing translation for 'onlineSavingsMsg'
Learn More
2-Cyanobenzeneboronic acid 1,3-propanediol ester, Thermo Scientific™ Product Code.: 15489895

2-Cyanobenzeneboronic acid 1,3-propanediol ester, Thermo Scientific™

Product Code. 15489895
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15489895

Brand: Thermo Scientific Alfa Aesar H52547.MD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172732-52-4
Molecular Formula C10H10BNO2
Molecular Weight (g/mol) 187.005
MDL Number MFCD04039012
InChI Key REQZFVYFYAZUMG-UHFFFAOYSA-N
Synonym 2-1,3,2-dioxaborinan-2-yl benzonitrile, 2-cyanophenylboronic acid 1,3-propanediol ester, 2-2-cyanophenyl-1,3,2-dioxaborinane, 2-cyanobenzeneboronic acid 1,3-propanediol ester, 2-cyanophenylboronic acid propanediol ester, 2-cyanophenylboronic acid, 1,3-propanediol cyclic ester, 2-1,3,2-dioxaborinane-2-yl benzonitrile, 2-1,3,2-dioxaborinan-2-yl-benzonitrile, benzonitrile, 2-1,3,2-dioxaborinan-2-yl, 2-cyanophenylboronic acid,1,3-propanediol
PubChem CID 3529602
IUPAC Name 2-(1,3,2-dioxaborinan-2-yl)benzonitrile
SMILES B1(OCCCO1)C2=CC=CC=C2C#N

Specifications

Melting Point 42°C to 45°C
Quantity 250 mg
Formula Weight 187
Chemical Name or Material 2-Cyanobenzeneboronic acid 1,3-propanediol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Cyanobenzeneboronic acid 1,3-propanediol ester, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.