missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Cyanoethyl)triethoxysilane, 97%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Specifications
Specifications
| Boiling Point | 224°C to 225°C |
| Quantity | 100 g |
| Beilstein | 1776392 |
| Flash Point | 100°C (212°F) |
| Solubility Information | Hydrolyzes in water. |
| Formula Weight | 217.34 |
| Percent Purity | 97% |
| Refractive Index | 1.413 |
| Sensitivity | Moisture sensitive |
| Density | 0.98 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction