missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoethylhydrazine, 95%, Thermo Scientific Chemicals

Product Code. 11410748
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanoethylhydrazine is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-07-1
Molecular Formula C3H7N3
Molecular Weight (g/mol) 85.11
MDL Number MFCD00043620
InChI Key VIHRIIARIFUQLC-UHFFFAOYSA-N
Synonym 2-cyanoethylhydrazine, beta-cyanoethylhydrazine, 2-cyanoethyl hydrazine, cyanoethylhydrazine, unii-l6h109c98b, 3-hydrazinopropionitrile, propanenitrile, 3-hydrazino, 3-hydrazinopropanenitrile
PubChem CID 136177
IUPAC Name 3-hydrazinylpropanenitrile
SMILES C(CNN)C#N

Specifications

Density 1.053
Boiling Point 76°C to 79°C (0.5 mmHg)
Flash Point 109°C (228°F)
Linear Formula NCCH2CH2NHNH2
Refractive Index 1.477
Quantity 1 g
UN Number UN2927
Beilstein 773834
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 85.11
Percent Purity 95%
Chemical Name or Material 2-Cyanoethylhydrazine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'