missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoethylhydrazine, 98%, Thermo Scientific Chemicals

Product Code. 10113441
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-07-1
Molecular Formula C3H7N3
Molecular Weight (g/mol) 85.1
MDL Number MFCD00043620
InChI Key VIHRIIARIFUQLC-UHFFFAOYSA-N
Synonym 2-cyanoethylhydrazine, beta-cyanoethylhydrazine, 2-cyanoethyl hydrazine, cyanoethylhydrazine, unii-l6h109c98b, 3-hydrazinopropionitrile, propanenitrile, 3-hydrazino, 3-hydrazinopropanenitrile
PubChem CID 136177
IUPAC Name 3-hydrazinylpropanenitrile
SMILES C(CNN)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.0530g/mL
Boiling Point 76°C to 79°C (0.5 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2NHCH2CH2CN
Refractive Index 1.4740 to 1.476
Quantity 1 g
Specific Gravity 1.053
Formula Weight 85.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Cyanoethylhydrazine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Cyanoethylhydrazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.