missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohepten-1-one, 90%, Thermo Scientific Chemicals

£62.30 - £231.00

Chemical Identifiers

CAS 1121-66-0
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
InChI Key WZCRDVTWUYLPTR-UHFFFAOYSA-N
Synonym 2-cyclohepten-1-one, cyclohept-2-enone, tropilene, 2-cycloheptenone, cycloheptenone, z-cyclohept-2-enone, cyclohept-2-eneone, cyclohept-en-1-one, acmc-1bphl, 2-cyclohepten-1-one, z
PubChem CID 70723
IUPAC Name cyclohept-2-en-1-one
SMILES C1CCC(=O)C=CC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15463534
View Documents
Thermo Scientific Acros
171070010
1 g
£62.30
1g
Please to purchase this item. Need a web account? Register with us today!
15595046
View Documents
Thermo Scientific Acros
171070050
5 g
£231.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1121-66-0
110.16
2-cyclohepten-1-one, cyclohept-2-enone, tropilene, 2-cycloheptenone, cycloheptenone, z-cyclohept-2-enone, cyclohept-2-eneone, cyclohept-en-1-one, acmc-1bphl, 2-cyclohepten-1-one, z
cyclohept-2-en-1-one
C7H10O
WZCRDVTWUYLPTR-UHFFFAOYSA-N
70723
C1CCC(=O)C=CC1

Specifications

1121-66-0
189.0°C
Authentic
Glass Bottle
C7H10(=O)
0.99
WZCRDVTWUYLPTR-UHFFFAOYSA-N
cyclohept-2-en-1-one
70723
90%
0.9900g/mL
68°C
89% min. (GC)
C7H10O
1.495
2-cyclohepten-1-one, cyclohept-2-enone, tropilene, 2-cycloheptenone, cycloheptenone, z-cyclohept-2-enone, cyclohept-2-eneone, cyclohept-en-1-one, acmc-1bphl, 2-cyclohepten-1-one, z
C1CCC(=O)C=CC1
110.16
110.16
2-Cyclohepten-1-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 214-334-1

Product Identifier
  • 2-Cyclohepten-1-one, 90%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only