Learn More
2-Cyclohexen-1-one, 97%, Thermo Scientific Chemicals
£37.00 - £395.00
Chemical Identifiers
CAS | 930-68-7 |
---|---|
Molecular Formula | C6H8O |
Molecular Weight (g/mol) | 96.13 |
MDL Number | MFCD00001577 |
InChI Key | FWFSEYBSWVRWGL-UHFFFAOYSA-N |
Synonym | 2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one |
PubChem CID | 13594 |
ChEBI | CHEBI:15977 |
IUPAC Name | cyclohex-2-en-1-one |
SMILES | O=C1CCCC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11472777
|
Thermo Scientific Alfa Aesar
A15435.14 |
25 g |
£37.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11482777
|
Thermo Scientific Alfa Aesar
A15435.22 |
100 g |
£98.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492777
|
Thermo Scientific Alfa Aesar
A15435.36 |
500 g |
£395.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Cyclohexen-1-one is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Cyclohexen-1-one is used as intermediates.
Solubility
Soluble in water, alcohols, benzene and acetone.
Notes
Store away from oxidizing agents and strong bases. Incompatible with reducing agents and bases.
Chemical Identifiers
930-68-7 | |
96.13 | |
FWFSEYBSWVRWGL-UHFFFAOYSA-N | |
13594 | |
cyclohex-2-en-1-one |
C6H8O | |
MFCD00001577 | |
2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one | |
CHEBI:15977 | |
O=C1CCCC=C1 |
Specifications
930-68-7 | |
0.993 | |
56°C (132°F) | |
1.488 | |
UN2929 | |
2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one | |
FWFSEYBSWVRWGL-UHFFFAOYSA-N | |
cyclohex-2-en-1-one | |
13594 | |
96.13 | |
2-Cyclohexen-1-one |
-53°C | |
167°C to 169°C | |
C6H8O | |
MFCD00001577 | |
1280477 | |
Soluble in water,alcohols,benzene and acetone. | |
O=C1CCCC=C1 | |
96.13 | |
CHEBI:15977 | |
97% |
Safety and Handling
GHS H Statement
H301-H310-H330-H226-H319
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapour.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P362-P374-P380-P501c
H226-H301-H310+H330-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 213-223-5
missing translation for 'rtecsNumber' : GW7000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only