missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethylboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

£46.10 - £134.00

Chemical Identifiers

CAS 167692-95-7
Molecular Formula C14H27BO2
Molecular Weight (g/mol) 238.178
MDL Number MFCD12546193
InChI Key LDKDGYZAIIKCGK-UHFFFAOYSA-N
Synonym 2-cyclohexylethylboronic acid pinacol ester, 2-2-cyclohexylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-cyclohexylethylboronicacid
PubChem CID 15259990
IUPAC Name 2-(2-cyclohexylethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2CCCCC2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424047
View Documents
Thermo Scientific Alfa Aesar
H31156.03
1 g
£46.10
1g
Please to purchase this item. Need a web account? Register with us today!
15434047
View Documents
Thermo Scientific Alfa Aesar
H31156.06
5 g
£134.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

167692-95-7
238.178
LDKDGYZAIIKCGK-UHFFFAOYSA-N
15259990
B1(OC(C(O1)(C)C)(C)C)CCC2CCCCC2
C14H27BO2
MFCD12546193
2-cyclohexylethylboronic acid pinacol ester, 2-2-cyclohexylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-cyclohexylethylboronicacid
2-(2-cyclohexylethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

167692-95-7
MFCD12546193
LDKDGYZAIIKCGK-UHFFFAOYSA-N
2-(2-cyclohexylethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
15259990
96%
C14H27BO2
2-cyclohexylethylboronic acid pinacol ester, 2-2-cyclohexylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-cyclohexylethylboronicacid
B1(OC(C(O1)(C)C)(C)C)CCC2CCCCC2
238.178
238.18
2-Cyclohexylethylboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.