missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexyloxypyridine-5-carboxaldehyde, 97%, Thermo Scientific™

£136.00 - £534.00

Chemical Identifiers

CAS 916792-16-0
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD08060944
InChI Key KTDSNAKFASLMHF-UHFFFAOYSA-N
Synonym 2-cyclohexyloxypyridine-5-carboxaldehyde, 6-cyclohexyloxy nicotinaldehyde, 6-cyclohexyloxy pyridine-3-carbaldehyde, 6-cyclohexyl-oxy pyridine-3-carbaldehyde
PubChem CID 42553152
IUPAC Name 6-cyclohexyloxypyridine-3-carbaldehyde
SMILES C1CCC(CC1)OC2=NC=C(C=C2)C=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11434590
View Documents
Thermo Scientific Alfa Aesar
H50129.03
1 g
£136.00
1g
Please to purchase this item. Need a web account? Register with us today!
11444590
View Documents
Thermo Scientific Alfa Aesar
H50129.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

916792-16-0
205.257
KTDSNAKFASLMHF-UHFFFAOYSA-N
42553152
C1CCC(CC1)OC2=NC=C(C=C2)C=O
C12H15NO2
MFCD08060944
2-cyclohexyloxypyridine-5-carboxaldehyde, 6-cyclohexyloxy nicotinaldehyde, 6-cyclohexyloxy pyridine-3-carbaldehyde, 6-cyclohexyl-oxy pyridine-3-carbaldehyde
6-cyclohexyloxypyridine-3-carbaldehyde

Specifications

916792-16-0
MFCD08060944
2-cyclohexyloxypyridine-5-carboxaldehyde, 6-cyclohexyloxy nicotinaldehyde, 6-cyclohexyloxy pyridine-3-carbaldehyde, 6-cyclohexyl-oxy pyridine-3-carbaldehyde
C1CCC(CC1)OC2=NC=C(C=C2)C=O
205.257
205.26
2-Cyclohexyloxypyridine-5-carboxaldehyde
C12H15NO2
Air and light sensitive
KTDSNAKFASLMHF-UHFFFAOYSA-N
6-cyclohexyloxypyridine-3-carbaldehyde
42553152
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications