Learn More
Cytiva 2-D Protein Extraction Buffer Trial Kit
Provides a convenient way to prepare high-quality protein lysates
Brand: Cytiva 28-9435-22
Code : 50
Additional Details : Weight : 0.25100kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Includes:
- 2-D Protein extraction buffer-I (Urea, NP-40)
- 2-D Protein extraction buffer-II (Urea, CHAPS)
- 2-D Protein extraction buffer-III (Urea, Thiourea, ASB-16, CHAPS)
- 2-D Protein extraction buffer-IV (Urea, Thiourea, SB 3-10, CHAPS)
- 2-D Protein extraction buffer-V (Urea, Thiourea, CHAPS)
- 2-D Protein extraction buffer-VI (Urea, Thiourea, NDSB 201, CHAPS)
- One 7mL DILUENT-I
- One 14mL DILUENT-II
- Two 12mL DILUENT-III
Description
Efficient cell lysis and protein extraction are main steps for achieving high-quality results in downstream applications.
- Buffers are designed to produce high spot resolution for 2-D gel analysis
- Six buffers are available as dry powder, which is rehydrated with the provided DILUENT
- Optimal buffer will depend on sample nature
- Allows to evaluate each extraction buffer to find the most suitable buffer for sample
Order Info
Complete packsize: 50mL
Specifications
White | |
Solid and Liquid | |
7 mL Diluent-I, 14 mL Diluent-II, 2 x 12 mL Diluent-III |
2-D Protein Extraction Buffer | |
|
|
2° to 8°C |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 2
- Carcinogenicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H302-Harmful if swallowed.
- H361d-Suspected of damaging the unborn child.
- H351-Suspected of causing cancer.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P405-Store locked up.
- P501-Dispose of contents/container to .