missing translation for 'onlineSavingsMsg'
Learn More

Cytiva 2-D Protein Extraction Buffers

Designed to produce high spot resolution for 2-D gel analysis

Brand:  Cytiva 28-9435-26

Product Code. 10076034

This item is not returnable. View return policy

Description

Description

  • Convenient way to prepare quality protein lysates
  • Ready to mix buffers simplifies protein extraction preparation of cells and tissues
  • Efficient and reproducible total protein extraction with yield
  • Can also be used prior to 1-D electrophoresis, or for rehydrating IPG strips to 2-D electrophoresis
  • Each package contains sufficient amounts of reagents to prepare 50mL buffer

Order Info

Buffer is supplied as a dry powder that is rehydrated with the provided diluent.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Contains <8M Urea, <5M Thiourea, <10% CHAPS
50 mL
15° to 30°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H361d-Suspected of damaging the unborn child.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers