missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-glucose, 98%

Product Code. 11476740
Click to view available options
Quantity:
1 g
5 g
25 g

2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-17-6
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00151328
InChI Key PMMURAAUARKVCB-PHUJZJCSNA-N
Synonym deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose
PubChem CID 17751002
IUPAC Name (3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O

Specifications

Melting Point 147°C to 152°C
Quantity 25 g
Beilstein 1723331
Merck Index 14,2904
Solubility Information Soluble in water.
Optical Rotation +47° (c=0.5 in Water, 45h)
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material 2-Deoxy-D-glucose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'