missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-ribose, 99%

Product Code. p-7081833
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 533-67-5 | C5H10O4 | 134.13 g/mol

2-Deoxy-D-ribose is used as a precursor to deoxyribonucleic acid. It induces apoptosis by inhibiting the synthesis and increasing the efflux of glutathione. Further, it is used in the synthesis of optically active dipyrrolyl alkanols from pyrroles on the surface of montmorillonite KSF clay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-ribose is used as a precursor to deoxyribonucleic acid. It induces apoptosis by inhibiting the synthesis and increasing the efflux of glutathione. Further, it is used in the synthesis of optically active dipyrrolyl alkanols from pyrroles on the surface of montmorillonite KSF clay.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-67-5
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
MDL Number MFCD00135904
InChI Key PDWIQYODPROSQH-GLULIZFNNA-N
Synonym 2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
PubChem CID 5460005
ChEBI CHEBI:28816
IUPAC Name (3S,4R)-3,4,5-trihydroxypentanal
SMILES OC[C@H]1OC(O)C[C@@H]1O

Specifications

Melting Point 76°C to 85°C
Quantity 1 g
Beilstein 1721978
Sensitivity Hygroscopic
Merck Index 14,2908
Solubility Information Soluble in water.
Optical Rotation −56° (c=1 in water, 24h)
Formula Weight 134.13
Percent Purity 99%
Chemical Name or Material 2-Deoxy-D-ribose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 2-Deoxy-D-ribose, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.