missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxycytidine-5'-triphosphate disodium salt, Thermo Scientific™

Product Code. 15437039
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
This item is not returnable. View return policy

Product Code. 15437039

Brand: Thermo Scientific Alfa Aesar J65686.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2′-Deoxycytidine 5′-triphosphate disodium salt is a nucleoside triphosphatase used for DNA synthesis with DNA polymerases and reverse transcriptases. 2′-Deoxycytidine 5′-triphosphate disodium salt is one of the nucleoside triphosphates used for DNA polymerase driven reactions such as polymerase chain reaction, DNA sequencing and other molecular biology methods.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102783-51-7
Molecular Formula C9H16N3Na2O13P3
Molecular Weight (g/mol) 513.14
MDL Number MFCD00084683,MFCD00080336
InChI Key QGRIWAGEUGZHQH-UHFFFAOYNA-N
Synonym 5'-dCTP Na2; 2'-Deoxy-5'-triphosphocytidine disodium salt
PubChem CID 131636617
IUPAC Name disodium;(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-2-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolan-3-olate
SMILES [Na].[Na].NC1=NC(=O)N(C=C1)C1CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O1

Specifications

Color White
Quantity 25 mg
Formula Weight 511.12
Physical Form Powder
Chemical Name or Material 2'-Deoxycytidine-5'-triphosphate disodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.