missing translation for 'onlineSavingsMsg'
Learn More
2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%, Thermo Scientific Chemicals Product Code.: 11408097

2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11408097
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11408097

Brand: Thermo Scientific Alfa Aesar A17485.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A sodium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-05-8
Molecular Formula C13H21ClN2O2
Molecular Weight (g/mol) 272.77
MDL Number MFCD00013000
InChI Key HCBIBCJNVBAKAB-UHFFFAOYSA-N
Synonym procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor
PubChem CID 5795
ChEBI CHEBI:8431
SMILES [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 155°C to 157°C
Boiling Point 195°C to 196°C (17 mmHg)
Odor Odorless
Quantity 250 g
UN Number UN2811
Sensitivity Light sensitive
Merck Index 14,7757
IUPAC Name hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride
Formula Weight 272.78
Percent Purity 99%
Chemical Name or Material 2-Diethylaminoethyl 4-aminobenzoate hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.