missing translation for 'onlineSavingsMsg'
Learn More
2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF, Thermo Scientific Chemicals Product Code.: 15495746

2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF, Thermo Scientific Chemicals

Product Code. 15495746
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15495746

Brand: Thermo Scientific Alfa Aesar H60435.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1053657-14-9
Molecular Formula C8H20NP
Molecular Weight (g/mol) 161.23
MDL Number MFCD11044863
InChI Key QUVUVMRUSAKOPB-UHFFFAOYSA-N
Synonym 2-diisopropylphosphino ethylamine, 2-di-i-propylphosphino ethylamine, 2-bis 1-methylethyl phosphino ethanamine, acmc-20aonc, 2-aminoethyldiisopropylphosphine, 2-di-isopropylphosphino ethanamine, 2-aminoethyl diisopropylphosphane, 2-di propan-2-yl phosphanyl ethan-1-amine, 2-aminoethyl bis propan-2-yl phosphane, 2-diisopropylphosphino ethylamine, kanata purity
PubChem CID 57552996
IUPAC Name 2-di(propan-2-yl)phosphanylethanamine
SMILES CC(C)P(CCN)C(C)C

Specifications

Linear Formula [(CH3)2CH]2PCH2CH2NH2
Quantity 25 g
UN Number UN2924
Sensitivity Air Sensitive
Formula Weight 161.22
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material 2-(Diisopropylphosphino)ethylamine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS24F041 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60435.14,H60435.06
Lot NumberK07Y013 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60435.14,H60435.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.