missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoisopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11419916
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11419916

Brand: Thermo Scientific Alfa Aesar B20882.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4584-49-0
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.066
MDL Number MFCD00012534
InChI Key OCWGRWAYARCRTQ-UHFFFAOYSA-N
Synonym 2-dimethylaminoisopropyl chloride hydrochloride, 2-chloro-1-dimethylamino propane hydrochloride, 2-chloro-n,n-dimethylpropan-1-amine hydrochloride, n,n-dimethyl-2-chloropropylamine hydrochloride, 2-chloropropyldimethylammonium chloride, 2-dimethylamino isopropyl chloride hydrochloride, 2-chloropropyldimethylamine hydrochloride, 2-chloropropyl-dimethylamine hydrochloride, 1-methyl-2-dimethylaminoethyl chloride hydrochloride, 2-chloropropyl dimethylamine hydrochloride
PubChem CID 94294
IUPAC Name 2-chloro-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CC(CN(C)C)Cl.Cl

Specifications

Melting Point 190°C to 193°C
Quantity 500 g
Beilstein 3670215
Sensitivity Hygroscopic
Formula Weight 158.07
Percent Purity 98%
Chemical Name or Material 2-Dimethylaminoisopropyl chloride hydrochloride
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.