missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoisopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

£19.80 - £54.70

Chemical Identifiers

CAS 4584-49-0
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.066
MDL Number MFCD00012534
InChI Key OCWGRWAYARCRTQ-UHFFFAOYSA-N
Synonym 2-dimethylaminoisopropyl chloride hydrochloride, 2-chloro-1-dimethylamino propane hydrochloride, 2-chloro-n,n-dimethylpropan-1-amine hydrochloride, n,n-dimethyl-2-chloropropylamine hydrochloride, 2-chloropropyldimethylammonium chloride, 2-dimethylamino isopropyl chloride hydrochloride, 2-chloropropyldimethylamine hydrochloride, 2-chloropropyl-dimethylamine hydrochloride, 1-methyl-2-dimethylaminoethyl chloride hydrochloride, 2-chloropropyl dimethylamine hydrochloride
PubChem CID 94294
IUPAC Name 2-chloro-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CC(CN(C)C)Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11409916
View Documents
Thermo Scientific Alfa Aesar
B20882.22
100 g
£19.80
100g
Please to purchase this item. Need a web account? Register with us today!
11419916
View Documents
Thermo Scientific Alfa Aesar
B20882.36
500 g
£54.70
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4584-49-0
158.066
OCWGRWAYARCRTQ-UHFFFAOYSA-N
94294
CC(CN(C)C)Cl.Cl
C5H13Cl2N
MFCD00012534
2-dimethylaminoisopropyl chloride hydrochloride, 2-chloro-1-dimethylamino propane hydrochloride, 2-chloro-n,n-dimethylpropan-1-amine hydrochloride, n,n-dimethyl-2-chloropropylamine hydrochloride, 2-chloropropyldimethylammonium chloride, 2-dimethylamino isopropyl chloride hydrochloride, 2-chloropropyldimethylamine hydrochloride, 2-chloropropyl-dimethylamine hydrochloride, 1-methyl-2-dimethylaminoethyl chloride hydrochloride, 2-chloropropyl dimethylamine hydrochloride
2-chloro-N,N-dimethylpropan-1-amine;hydrochloride

Specifications

4584-49-0
C5H13Cl2N
3670215
2-dimethylaminoisopropyl chloride hydrochloride, 2-chloro-1-dimethylamino propane hydrochloride, 2-chloro-n,n-dimethylpropan-1-amine hydrochloride, n,n-dimethyl-2-chloropropylamine hydrochloride, 2-chloropropyldimethylammonium chloride, 2-dimethylamino isopropyl chloride hydrochloride, 2-chloropropyldimethylamine hydrochloride, 2-chloropropyl-dimethylamine hydrochloride, 1-methyl-2-dimethylaminoethyl chloride hydrochloride, 2-chloropropyl dimethylamine hydrochloride
CC(CN(C)C)Cl.Cl
158.066
158.07
2-Dimethylaminoisopropyl chloride hydrochloride
190°C to 193°C
MFCD00012534
Hygroscopic
OCWGRWAYARCRTQ-UHFFFAOYSA-N
2-chloro-N,N-dimethylpropan-1-amine;hydrochloride
94294
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 224-971-7

missing translation for 'rtecsNumber' : TY1225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.