missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzonitrile, 98%, Thermo Scientific™

Product Code. 10453160
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6609-57-0
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00001789
InChI Key DXTLCLWOCYLDHL-UHFFFAOYSA-N
Synonym o-ethoxybenzonitrile, benzonitrile, ethoxy, 2-ethoxybenzenecarbonitrile, o-ethoxybenzontrle, acmc-20anwu, 2-ethoxy benzonitrile, 2-ethoxy-benzonitrile, pubchem4773, benzonitrile, 2-ethoxy, 2-ethoxybenzonitrile
PubChem CID 576922
IUPAC Name 2-ethoxybenzonitrile
SMILES CCOC1=CC=CC=C1C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5°C
Color Colorless to Yellow
Boiling Point 45°C (0.5 Torr)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 147.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethoxybenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2-Ethoxybenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'