missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxybenzonitrile, 98%, Thermo Scientific™
£27.84 - £102.31
Chemical Identifiers
CAS | 6609-57-0 |
---|---|
Molecular Formula | C9H9NO |
Molecular Weight (g/mol) | 147.18 |
MDL Number | MFCD00001789 |
InChI Key | DXTLCLWOCYLDHL-UHFFFAOYSA-N |
Synonym | o-ethoxybenzonitrile, benzonitrile, ethoxy, 2-ethoxybenzenecarbonitrile, o-ethoxybenzontrle, acmc-20anwu, 2-ethoxy benzonitrile, 2-ethoxy-benzonitrile, pubchem4773, benzonitrile, 2-ethoxy, 2-ethoxybenzonitrile |
PubChem CID | 576922 |
IUPAC Name | 2-ethoxybenzonitrile |
SMILES | CCOC1=CC=CC=C1C#N |
Description
Chemical Identifiers
6609-57-0 | |
147.18 | |
DXTLCLWOCYLDHL-UHFFFAOYSA-N | |
576922 | |
CCOC1=CC=CC=C1C#N |
C9H9NO | |
MFCD00001789 | |
o-ethoxybenzonitrile, benzonitrile, ethoxy, 2-ethoxybenzenecarbonitrile, o-ethoxybenzontrle, acmc-20anwu, 2-ethoxy benzonitrile, 2-ethoxy-benzonitrile, pubchem4773, benzonitrile, 2-ethoxy, 2-ethoxybenzonitrile | |
2-ethoxybenzonitrile |
Specifications
6609-57-0 | |
100.0 | |
Colorless to Yellow | |
113°C | |
97.5% min. (GC) | |
C9H9NO | |
o-ethoxybenzonitrile, benzonitrile, ethoxy, 2-ethoxybenzenecarbonitrile, o-ethoxybenzontrle, acmc-20anwu, 2-ethoxy benzonitrile, 2-ethoxy-benzonitrile, pubchem4773, benzonitrile, 2-ethoxy, 2-ethoxybenzonitrile | |
CCOC1=CC=CC=C1C#N | |
147.18 | |
147.18 | |
Liquid |
97.5 | |
5°C | |
45°C (0.5 Torr) | |
Authentic | |
Glass Bottle | |
MFCD00001789 | |
DXTLCLWOCYLDHL-UHFFFAOYSA-N | |
2-ethoxybenzonitrile | |
576922 | |
98% | |
2-Ethoxybenzonitrile |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Ethoxybenzonitrile
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only