missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Ethoxybenzoyl chloride, 98%
Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
Boiling Point | 114°C to 118°C (5 mmHg) |
Quantity | 25 g |
UN Number | UN3265 |
Synonym | o-ethoxybenzoyl chloride, benzoyl chloride, 2-ethoxy, 2-ethoxybenzoylchloride, acmc-209jrt, 2-ethoxy-benzoyl chloride, benzoyl chloride,2-ethoxy, 2-ethoxybenzoic acid chloride, ksc491c5l, 2-ethoxybenzoyl chloride |
Formula Weight | 184.62 |
Percent Purity | 98% |
Chemical Name or Material | 2-Ethoxybenzoyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'