missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxythiobenzamide, 97%, Thermo Scientific™

Product Code. 15466167
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 725702-35-2
Molecular Formula C9H11NOS
Molecular Weight (g/mol) 181.253
MDL Number MFCD06090837
InChI Key FUQRHZQJAIHJQB-UHFFFAOYSA-N
Synonym 2-ethoxybenzene-1-carbothioamide, 2-ethoxybenzothioamide, 2-ethoxythiobenzamide, benzenecarbothioamide,2-ethoxy, amino 2-ethoxyphenyl methane-1-thione
PubChem CID 7745241
IUPAC Name 2-ethoxybenzenecarbothioamide
SMILES CCOC1=CC=CC=C1C(=S)N

Specifications

Melting Point 113°C to 119°C
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 181.25
Percent Purity 97%
Chemical Name or Material 2-Ethoxythiobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'