missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1,3-cyclopentanedione, 98%, Thermo Scientific Chemicals

Product Code. 11469736
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-36-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00044191
InChI Key YDFBIBUYOUFJMR-UHFFFAOYSA-N
Synonym 2-ethyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-ethyl, unii-91a82te3os, acmc-209poa, 2-ethyl-cyclopentan-1,3-dione, 2-ethyl-cyclopentane-1,3-dione
PubChem CID 69978
IUPAC Name 2-ethylcyclopentane-1,3-dione
SMILES CCC1C(=O)CCC1=O

Specifications

Melting Point 168°C to 170°C
Quantity 5 g
Beilstein 1860068
Formula Weight 126.16
Percent Purity 99%
Chemical Name or Material 2-Ethyl-1,3-cyclopentanedione

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'