missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4,5-dimethyloxazole, 98%, Thermo Scientific™

£23.20 - £66.30

Chemical Identifiers

CAS 53833-30-0
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD00036664
InChI Key LCYOFVYHDBWYSI-UHFFFAOYSA-N
PubChem CID 62069
IUPAC Name 2-ethyl-4,5-dimethyl-1,3-oxazole
SMILES CCC1=NC(=C(O1)C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317349
View Documents
Thermo Scientific Alfa Aesar
H27470.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11327349
View Documents
Thermo Scientific Alfa Aesar
H27470.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

53833-30-0
125.171
LCYOFVYHDBWYSI-UHFFFAOYSA-N
2-ethyl-4,5-dimethyl-1,3-oxazole
C7H11NO
MFCD00036664
62069
CCC1=NC(=C(O1)C)C

Specifications

53833-30-0
1.511
2-ethyl-4,5-dimethyloxazole, oxazole, 2-ethyl-4,5-dimethyl, 2-ethyl-4,5-dimethyl oxazole, 4,5-dimethyl-2-ethyloxazole, unii-0w53s23ng4, fema no. 3672, 2-ethyl-4,5-dimethyl-oxazole, oxazole,2-ethyl-4,5-dimethyl, #
CCC1=NC(=C(O1)C)C
125.171
125.18
2-Ethyl-4,5-dimethyloxazole
C7H11NO
MFCD00036664
LCYOFVYHDBWYSI-UHFFFAOYSA-N
2-ethyl-4,5-dimethyl-1,3-oxazole
62069
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 258-815-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications