missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylamino-3-methoxypyridine, 99%, Thermo Scientific™

Product Code. 11449810
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 912761-77-4
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.20
MDL Number MFCD08061118
InChI Key ZHSBBKCVTLHSHQ-UHFFFAOYSA-N
Synonym 2-ethylamino-3-methoxypyridine, acmc-20anwe, ethyl 3-methoxy 2-pyridyl amine, n-ethyl-3-methoxy-2-pyridinamine, ethyl-3-methoxy-pyridin-2-yl-amine
PubChem CID 28054519
IUPAC Name N-ethyl-3-methoxypyridin-2-amine
SMILES CCNC1=C(OC)C=CC=N1

Specifications

Quantity 1 g
Formula Weight 152.2
Percent Purity 99%
Chemical Name or Material 2-Ethylamino-3-methoxypyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'