missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals

Product Code. 10559600
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-57-5
MDL Number MFCD00002675
InChI Key OBETXYAYXDNJHR-UHFFFAOYSA-N
Synonym 2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid
PubChem CID 8697
IUPAC Name 2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -59.0°C
Density 0.9060g/mL
Boiling Point 228.0°C
Flash Point 114°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H16O2
Linear Formula CH3(CH2)3CH(C2H5)CO2H
Refractive Index 1.4240 to 1.4260
Quantity 500 g
Beilstein 02,349
Specific Gravity 0.906
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in ether
Molecular Weight (g/mol) 144.21
Viscosity 7.7 mPa.s (20°C)
Formula Weight 144.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexanoic acid
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'