Learn More
2-Ethylhexylamine, 99%, Thermo Scientific Chemicals
£22.50 - £161.00
Chemical Identifiers
CAS | 104-75-6 |
---|---|
Molecular Formula | C8H19N |
Molecular Weight (g/mol) | 129.25 |
MDL Number | MFCD00008148 |
InChI Key | LTHNHFOGQMKPOV-UHFFFAOYSA-N |
Synonym | 2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame |
PubChem CID | 7719 |
IUPAC Name | 2-ethylhexan-1-amine |
SMILES | CCCCC(CC)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10678504
|
Thermo Scientific Acros
271661000 |
100 mL | Glass bottle |
£22.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10378810
|
Thermo Scientific Acros
271662500 |
250 mL | Glass bottle |
£23.30
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10195110
|
Thermo Scientific Acros
271660010 |
1 L | Glass bottle |
£65.20
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13418498
|
Thermo Scientific Acros
271660025 |
2.5 L | Glass Bottle |
£161.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
104-75-6 | |
129.25 | |
LTHNHFOGQMKPOV-UHFFFAOYSA-N | |
7719 | |
CCCCC(CC)CN |
C8H19N | |
MFCD00008148 | |
2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame | |
2-ethylhexan-1-amine |
Specifications
104-75-6 | |
100.0 | |
Colorless | |
169.0°C | |
Authentic | |
C8H19N | |
1.4295 to 1.4315 | |
04,388 | |
2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame | |
LTHNHFOGQMKPOV-UHFFFAOYSA-N | |
2-ethylhexan-1-amine | |
7719 | |
129.25 | |
Liquid |
98.5 | |
-76.0°C | |
0.7800g/mL | |
50°C | |
99% | |
CH3(CH2)3CH(C2H5)CH2NH2 | |
MFCD00008148 | |
0.78 | |
Solubility in water: 2.5g/L (20 c). | |
CCCCC(CC)CN | |
129.25 | |
1.1 mPa.s (20°C) | |
99% | |
2-Ethylhexylamine |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-233-8
- 2-Ethylhexylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H312-Harmful in contact with skin.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only