missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylpyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.
Solubility
Miscible with water (42 g/L at20°C).
Notes
Store at room temperature. Keep away from strong oxidizing agents.
2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.
Solubility
Miscible with water (42 g/L at20°C).
Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications
Melting Point | -63°C |
Density | 0.937 |
pH | 9 to 10 |
Boiling Point | 149°C |
Flash Point | 37°C (98°F) |
Odor | Unpleasant |
Refractive Index | 1.497 |
Quantity | 100 g |
UN Number | UN1993 |
Beilstein | 106480 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'