missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro[1,1-biphenyl]-4-carboxylic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Linear Formula | FC6H4C6H4CO2H |
Quantity | 1 g |
IUPAC Name | 4-(2-fluorophenyl)benzoic acid |
Formula Weight | 216.21 |
Percent Purity | 95% |
Chemical Name or Material | 2'-Fluoro[1, 1'-biphenyl]-4-carboxylic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Fluoro[1,1-biphenyl]-4-carboxylic acid, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction