missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11315475
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352303-67-4
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD02094728
InChI Key JCKZNMSBFBPDPM-UHFFFAOYSA-N
Synonym 2-fluoro-3-methoxyphenyl boronic acid, 2-fluoro-3-methoxybenzeneboronic acid, 2-fluoro-3-methoxyphenyl boranediol, 2-fluoro-3-methoxyphenylboronicacid, boronic acid, 2-fluoro-3-methoxyphenyl, pubchem1848, 2-fluoro-3-methoxy-phenyl boronic acid, acmc-1af04, 2-fluoro-3-methoxy-phenylboronic acid
PubChem CID 4985744
IUPAC Name (2-fluoro-3-methoxyphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)OC)F)(O)O

Specifications

Quantity 1 g
Beilstein 9191510
Formula Weight 169.95
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-methoxybenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'