missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 15428045
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15428045

Brand: Thermo Scientific Alfa Aesar H32292.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride, is used as a pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Moisture&water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017778-34-5
Molecular Formula C9H2ClF7O
Molecular Weight (g/mol) 294.553
MDL Number MFCD09258670
InChI Key RBJVFNYOFYHETI-UHFFFAOYSA-N
Synonym 2-fluoro-4,6-bis trifluoromethyl benzoyl chloride
PubChem CID 46737582
IUPAC Name 2-fluoro-4,6-bis(trifluoromethyl)benzoyl chloride
SMILES C1=C(C=C(C(=C1C(F)(F)F)C(=O)Cl)F)C(F)(F)F

Specifications

Refractive Index 1.4146
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 294.55
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.