missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitroanisole, 95%, Thermo Scientific™

Product Code. 15414367
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-93-6
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.127
MDL Number MFCD00061095
InChI Key XGMVTXUXZUPGGY-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitroanisole, 3-fluoro-4-methoxynitrobenzene, benzene, 2-fluoro-1-methoxy-4-nitro, 2-fluoro-1-methoxy-4-nitro-benzene, pharmabridge p-3000, 2-fluor-1-methoxy-4-nitrobenzol, pubchem2145, 2-fluoro-4-nitroanisol, 4-nitro-2-fluoroanisol, acmc-1apvd
PubChem CID 223104
IUPAC Name 2-fluoro-1-methoxy-4-nitrobenzene
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])F

Specifications

Melting Point 102°C to 106°C
Quantity 25 g
Formula Weight 171.13
Percent Purity 95%
Chemical Name or Material 2-Fluoro-4-nitroanisole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'