missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 10255114
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 126917-10-0
Molecular Formula C8H3ClF4O
Molecular Weight (g/mol) 226.56
MDL Number MFCD00061155
InChI Key OOAHPLWBUUTFMV-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl benzoyl chloride, 2-fluoro-4-trifluoromethyl benzoylchloride, benzoyl chloride, 2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-benzoyl chloride, benzoyl chloride,2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-1-benzenecarbonyl chloride, alpha,alpha,alpha,2-tetrafluoro-p-toluoyl chloride, acmc-1c5cr, 2-fluoro-4-trifluoromethyl-benzoylchloride, 4-trifluoromethyl-2-fluorobenzoyl chloride
PubChem CID 145580
IUPAC Name 2-fluoro-4-(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=C(C=C1C(F)(F)F)F)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.5000g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(CF3)COCl
Refractive Index 1.4650 to 1.4670 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.5
Formula Weight 226.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-4-(trifluoromethyl)benzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.